missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PITC (Edman's Reagent) (phenylisothiocyanate)

Product Code. 10044944 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10044944 10 x 1 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10044944 Supplier Thermo Scientific™ Supplier No. TS26922

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Phenylisothiocyanate, an amine-reactive, colored compound (Amax 254nm; UV) for pre-column derivatization of amino acids for reverse-phase HPLC.

Thermo Scientific™ Pierce PITC (phenylisothiocyanate) is a high-purity reagent for pre-column quantitative derivatization of amino acids by reverse-phase HPLC.

Highlights:

  • Also known as Edman's Reagent
  • Reacts readily with amino acids in 5 to 10 minutes at room temperature
  • Resulting phenylthiocarbamyl derivatives can be separated and quantified in 30 minutes using reverse-phase HPLC to produce stable products with all amino acids including proline

Ordering Alerts:

X in the ordering table indicates that hazardous shipping charges apply.

TRUSTED_SUSTAINABILITY

Specifications

Product Type PITC
Chemical Reactivity Amine
Content And Storage Upon receipt store at room temperature.
Label or Dye Other Label or Dye
Quantity 10 x 1 mL
Form Liquid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenylisothiocyanate (PITC)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.