Learn More
Thermo Scientific™ Pierce™ His Protein Interaction Pull-Down Kit
Shop All Thermo Scientific ProductsDescripción
Thermo Scientific™ Pierce PolyHis Protein Interaction Pull-Down Kit purifies protein interaction complexes involving practically any user-supplied His-tagged fusion protein.
Highlights:
6xHis pull-down – purifies protein interactors of any His-tagged fusion protein
Complete kit – provides all components and detailed protocol for purifying protein:protein interactions
No special equipment needed – uses common laboratory equipment and reagents (e.g., microcentrifuge)
Convenient – microcentrifuge spin columns facilitate simple and efficient manipulation of cobalt IMAC agarose beads, including simple processing of multiple samples
Flexible – instructions include protocols for bait and prey proteins from different sources
Includes:
Cobalt chelate agarose resin, lysis buffer, TBS pack, imidazole solution for elution, microcentrifuge spin columns and collection tubes
Recommended for:
Discover a new protein:protein interaction from a cell lysate; Confirm a putative interaction from a cell lysate or with a previously purified protein; Extract protein:protein interaction information from in vitro transcription/translation lysates
Especificaciones
Especificaciones
| Content And Storage | Sufficient For: At least 25 pull-down reactions, each using 25 μL of resin • Cobalt Chelate Agarose Resin, 750 μL • Pull-Down Lysis Buffer, 250 mL • Imidazole Solution (4M), 5 mL • TBS Pack (makes 500 mL), 1 pack • Pierce Spin Columns and Accessories, 27 columns • Microcentrifuge Collection Tubes, 100 tubes Upon receipt store Pierce Lysis Buffer at room temperature. Store remaining kit components at 4°C. |
| Assay | Affinity Purification |
| For Use With (Equipment) | Microcentrifuge |
| Product Line | Pierce |
| Quantity | 25 reactions kit |
| For Use With (Application) | Pull-Down (Tag-Based) |
| Target | Not Target-Specific |
| Type | Pull-Down Kit |
| Sufficient For | 25 Reactions |
| Format | Kit |
Seguridad y manipulación
- Pierce His Protein Interaction Pull-Down Kit
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H360-May damage fertility or the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 1-Imidazole, ethanol, Sodium chloride
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.