missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Prestained Protein MW Marker

Product Code. 11583100 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 250 μL
8 x 250 μL
Unit Size:
Each
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11583100 2 x 250 μL Each
17166098 8 x 250 μL Pack of 4
15513197 8 x 250 μL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11583100 Supplier Thermo Scientific™ Supplier No. 26612
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120 kDa) for use as size standards in protein electrophoresis (SDS-PAGE).

Thermo Scientific™ Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120kDa) for use as size standards in protein electrophoresis (SDS-PAGE).

Thermo Scientific Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120 kDa) for use as size standards in protein electrophoresis (SDS-PAGE). The protein standards are supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Component proteins

  • Beta-galactosidase (120 kDa)
  • Bovine serum albumin (85 kDa)
  • Ovalbumin (50 kDa)
  • Cardonic anhydrase (35 kDa)
  • Beta-lactoglobulin (25 kDa)
  • Lysozyme (20 kDa)

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis.
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage Buffer: 62.5 mM Tris · H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% (w/v) SDS, 1 mM DTT, 1.5 mM NaN3 and 33% (v/v) glycerol.

Upon receipt store at -20°C. Product is shipped with an ice pack.
Color Blue
Detection Method Colorimetric
Number of Markers 6
Ready to Load Yes
Size Range 20 to 120 kDa
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 120, 85, 50, 35, 25, 20 kDa
Quantity 2 x 250 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
Product Line Pierce
Product Type Protein Molecular Weight (MW) Marker
System Type Western Blotting, SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.