Learn More
Description
Thermo Scientific™ Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
Thermo Scientific Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120 kDa) for use as size standards in protein electrophoresis (SDS-PAGE). The protein standards are supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Component proteins
- Beta-galactosidase (120 kDa)
- Bovine serum albumin (85 kDa)
- Ovalbumin (50 kDa)
- Cardonic anhydrase (35 kDa)
- Beta-lactoglobulin (25 kDa)
- Lysozyme (20 kDa)
Applications
- Monitoring protein migration during SDS-polyacrylamide gel electrophoresis.
- Monitoring protein transfer onto membranes after western blotting
- Sizing of proteins on SDS-polyacrylamide gels and western blots
Specifications
Specifications
| Content And Storage | Storage Buffer: 62.5 mM Tris · H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% (w/v) SDS, 1 mM DTT, 1.5 mM NaN3 and 33% (v/v) glycerol. Upon receipt store at -20°C. Product is shipped with an ice pack. |
| Color | Blue |
| Detection Method | Colorimetric |
| Number of Markers | 6 |
| Ready to Load | Yes |
| Size Range | 20 to 120 kDa |
| Molecular Weight (g/mol) | 120, 85, 50, 35, 25, 20 kDa |
| Quantity | 8 x 250 μL |
| Product Line | Pierce |
| Product Type | Protein Molecular Weight (MW) Marker |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.