missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Protease Inhibitor XL Capsules, EDTA-free
Shop All Thermo Scientific ProductsDescription
Includes
10 Capsules
- Broad spectrum formulation—contains AEBSF, bestatin, E-64 and pepstatin A for better protection versus PMSF
- Large volume format – one capsule is sufficient for 500mL of solution
- Convenient—ready-to-use, dissolves clearly into solution within 3 minutes
- Compatible—use directly with Pierce Protein Assays
- EDTA-free—formulated without EDTA, a metalloproteinase inhibitor
Specifications
Specifications
| Product Type | Protease Inhibitor Cocktail |
| Content And Storage | 1 Vial, Store at 2°C to 8°C |
| Shipping Condition | Room Temperature |
| Form | Solid |
| Product Line | Pierce |
| Sufficient For | 500 mL Solution |
| Inhibitors | Protease Inhibitor |
| Quantity | 10 Capsules |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?