missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bestatin Protease Inhibitor

Product Code. 10687894 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10687894 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10687894 Supplier Thermo Scientific™ Supplier No. 78433
  • ONLINE EXCLUSIVE PRICE
    286.00 €
    248.82 € / 10mg
    Save 37.18 €
    13% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Customize your protease inhibitor cocktail formulation with these small packages of individual protease inhibitor peptides and compounds.

Thermo Scientific™ Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.

The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A, and PMSF. Although Thermo Scientific Halt™ Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.

Highlights:

  • High-quality reagents in convenient and affordable package sizes
  • Purchase individually to prepare customized protease inhibitor cocktails
  • Supplement Halt Protease Inhibitor Single-Use Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Bestatin Protease Inhibitor
Product Type Inhibitor
Content And Storage Store between 2°C and 8°C
Form Lyophilized
Product Line Pierce
Reagent Type Protease Inhibitor
Inhibitors Protease Inhibitor
Quantity 10 mg

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.