missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AEBSF Protease Inhibitor

Product Code. 11844101 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11844101 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11844101 Supplier Thermo Scientific™ Supplier No. 78431
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Customize your protease inhibitor cocktail formulation with these small packages of individual protease inhibitor peptides and compounds.

Thermo Scientific™ Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.

The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A, and PMSF. Although Thermo Scientific Halt™ Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.

Highlights:

  • High-quality reagents in convenient and affordable package sizes
  • Purchase individually to prepare customized protease inhibitor cocktails
  • Supplement Halt Protease Inhibitor Single-Use Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce AEBSF Protease Inhibitor
Product Type Protease Inhibitor
Content And Storage Store between 2°C and 25°C
Form Lyophilized
Product Line Pierce
Reagent Type Protease Inhibitor
Inhibitors Protease Inhibitor
Quantity 100 mg
compliance-icons
Product Identifier
  • AEBSF Protease Inhibitor
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 4-(2-aminoethyl)-benzenesulfonyl fluoride, hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.