missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets,EDTA-free

Product Code. 16316532 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 Tablets
Includes:
1 Vial Inhibitor Tablets
1 Vial Inhibitor Tablets, EDTA Free
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Includes unitSize
16316532 2 Tablets 1 Vial Inhibitor Tablets Each
16326532 2 Tablets 1 Vial Inhibitor Tablets, EDTA Free Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.
Product Code. 16316532 Supplier Thermo Scientific™ Supplier No. A32960
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. Not for use in diagnostic procedures.

Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets broad-spectrum protease and pghospahatase inhibitor formulation provides complete protection from dephosphorylation and proteolytic degradation.

The Pierce Protease and Phosphatase Inhibitor Mini Tablets contain aprotinin, bestatin, E-64, EDTA, and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.

Highlights:
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
* EDTA is present only in Product No. A32959.

TRUSTED_SUSTAINABILITY

Specifications

Includes 1 Vial Inhibitor Tablets
Product Type Protease and Phosphatase Inhibitor
Content And Storage 2°C to 8°C
For Use With (Application) protein purification, western blotting
Sufficient For 1 Tablet is Sufficient for 10 mL
Quantity 2 Tablets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.