missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Proteinase K Powder (≥30 U/mg)

Product Code. 11826724 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11826724 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11826724 Supplier Thermo Scientific™ Supplier No. 17916
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cleave at the carboxyl-side of aliphatic, aromatic or hydrophobic residues and digest-inactivate DNase and RNase in nucleic acid purification using this protease.

Thermo Scientific™ Pierce Proteinase K is a protease that cleaves at the carboxyl side of aliphatic, aromatic or hydrophobic residues and is commonly used to digest and inactivate DNase and RNase during in nucleic acid purification.

Calcium is not essential to the function of proteinase K. Therefore, EDTA and other chelating agents do not interfere with the activity and may be used alongside proteinase K to inactivate calcium-dependent nucleases in DNA and RNA preparation.

Highlights:

  • Has a high activity that is stable across a wide range of pH and temperature conditions; suited to short digestion times
  • Activity is increased at elevated temperatures up to 65°C
  • RNase- and DNase-free

Ordering Alerts:

Supplied lyophilized.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Form Powder
Quantity 100 mg
Product Type Proteinase K
How do you recommend storing proteinase K?

We recommend storing it at 4°C.

compliance-icons compliance-icons
Product Identifier
  • Proteinase K Powder
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: proteases

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.