missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Renilla-Firefly Luciferase Dual Assay Kit

Product Code. p-4532170 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12201682 100 Reaction Kit Each
12201692 1000 Reaction Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12201682 Supplier Thermo Scientific™ Supplier No. 16185

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Simultaneously detect intracellular Renilla and Red Firefly luciferase activity in mammalian whole cell lysates.

The Thermo Scientific™ Pierce Renilla-Firefly Luciferase Dual Assay Kit provides the necessary reagents to simultaneously detect intracellular Renilla and Red Firefly luciferase activity in mammalian whole cell lysates.

The Pierce Renilla-Firefly Luciferase Dual Assay Kit is a highly sensitive assay that allows for the simultaneous detection of Green Renilla and Red Firefly luciferase activity. Green Renilla luciferase acts as an experimental reporter with constitutively active red firefly as a normalization control. This reporter-and-control combination enables simultaneous monitoring of experimental reporter and control luciferase activities in a single-read assay without the need for two-step addition of substrate reagents or quenching.

The assay working solution contains substrates for both luciferases, and the reactions occur simultaneously with flash-type kinetics. The resulting luminescent signals are spectrally resolvable using filters. In a single sample, researchers can assay transcriptional activity of regulatory elements, signal transduction pathways, and effects of activators or inhibitors.

Highlights

  • Simultaneous—concurrent, filter-based, wavelength-separated detection of two luciferase activities
  • Sensitive—measure Green Renilla and red firefly luciferase activities in the same sample
  • Fast—no quenching step required, unlike in traditional sequential dual assays
  • Multiplex—capable of quantitating two cellular activities in the same sample(s)

Requires

Green Renilla and Red Firefly luciferase reporters; filter sets at 525 nm±20 nm for Green Renilla Luc and 615 to 675 nm for Red Firefly Luc; luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers; injector for assessing more than 24 wells at a time

Recommended applications

Study two regulatory elements at the same time; monitor two signaling pathways simultaneously; enable studying more than one target per screen (e.g., off-target effects)

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Firefly Luciferase, Green Renilla Luciferase
Sufficient For 100 Dual Luciferase Assays
Quantity 100 Reaction Kit
Content And Storage Sufficient For: 100 dual-luciferase assays in microplate wells
• Renilla-Firefly Dual Assay Buffer, 5 mL (store at 4°C)
• 100X Coelenterazine, 50 μL (store at -80°C)
• D-Luciferin, 3 mg (store at 4°C)
• Luciferase Cell Lysis Buffer (2X), 6 mL (store at room temperature)

Upon receipt, store kit at -80°C, or store individual components as indicated above.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate Coelenterazine, D-Luciferin
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pierce Renilla-Firefly Luciferase Dual Assay Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.