missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Renilla Luciferase Glow Assay Kit

Product Code. 12182240 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12182240 1000 Reaction Kit Each
12172240 100 Reaction Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12182240 Supplier Thermo Scientific™ Supplier No. 16167
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Obtain bright and stable bioluminescence signal (half-life approx. 3 hour) in the presence of Renilla luciferase reporters.

The Thermo Scientific Pierce Renilla Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life approx. 3 hours), especially in the presence of Green Renilla luciferase reporter.

Features of the Renilla Luciferase Glow Assay Kit:

  • Sensitive—highly sensitive detection of Green Renilla luciferase activity
  • Stable—increased signal stability compared to flash assays
  • Simple—does not require a luminometer with injectors
  • Compatible—assay reagents compatible with other Renilla luciferases
  • Automation-friendly—amenable to high throughput assays
  • Convenient—contains a universal cell lysis buffer and optimized glow assay reagent
  • Safe—allows one to perform non-radioactive assays

This Pierce Luciferase Glow Assay Kit contains reagents for measuring the activity of Renilla luciferase in cell lysates. In the presence of luciferase, the glow reagent provides stable bioluminescent signal and is highly suited for luminometers without injectors or for batch processing of samples. The light output generated by the luciferase reaction can be correlated with the amount of luciferase protein produced which in turn is proportional to the promoter activity driving the luciferase expression. The glow assay reagents were optimized for best results with Thermo Scientific Green Renilla Luc Plasmids; however, this glow kit may be used to detect the activity of other Renilla luciferase derivatives that use coelenterazine as a substrate.

Includes:

Cell lysis buffer, reaction buffer and substrate

Requires:

Renilla luciferase and luminometer or other instrument capable of monitoring luminescence, such as the Varioskan ALF or the Varioskan LUX microplate readers.

Applications:

  • Promoter studies for analyzing cis- regulatory elements and trans-acting factors
  • Drug screening
  • siRNA and miRNA screening
  • Multiplexed assays to study off-target effects
  • Protein localization reporter assays
  • Signal transduction pathway analysis
  • RNA splicing studies

The Green Renilla luciferase is a 36kDa protein produced by a derivative of the wild type Renilla luciferase gene from the sea pansy, Renilla reniformis. Compared to the wild type luciferase, Green Renilla is more stable in serum and has an the emission spectrum that is shifted toward the green region. The protein provides extremely bright flash signal that decays rapidly. The Pierce Renilla Glow Assay Kit reagent effectively stabilizes this bright bioluminescence to yield a signal half-life greater than 3 hours, thus eliminating the need for a luminometer with injectors. The assay kit was optimized for best results with Thermo Scientific Green Renilla Luc Plasmids; however, this glow kit may be used to detect the activity of other Renilla luciferase derivatives that use coelenterazine as a substrate.

Related Products

Pierce™ Renilla-Firefly Luciferase Dual Assay Kit (Cat. No. 16185)

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Green Renilla Luciferase
Quantity 1000 Reaction Kit
Content And Storage Upon receipt store kit at -80°C or store individual components as indicated above.
• Renilla Glow Assay Buffer, 50 mL, store at 4°C
• Coelenterazine (100X), 0.5 mL, store at -80°C
• 2X Cell Lysis Buffer, 60 mL, store at room temperature
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer
Format 96-well Plate, 384-well Plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate Coelenterazine
Show More Show Less
I got a high background signal with a Gaussia/Cypridina/Renilla Luciferase Glow/Luciferase Flash assay. What went wrong?

Here are possible causes and solutions:

- Non-specific oxidation of substrate: Use less serum in the cell culture media; Note: Albumin can increase the auto-oxidation of Vargulin; Avoid repeated freezing and thawing of the sample.
- Control sample is contaminated: Use new sample; Change pipette tips after each well.

I am getting a high saturating signal with a Gaussia/Cypridina/Renilla Luciferase Glow/Luciferase Flash assay. What could have happened?

This could be due to high luciferase expression. Here are some suggestions:

  • Reduce incubation time before collecting samples. 
  • Decrease the integration time on the instrument.
  • Dilute the sample: for secreted Gaussia/Cypridina Luciferase, dilute the sample using media from the cell culture; for cell lysate, dilute the sample using lysis buffer 


  • Note: A low sample volume can increase assay variability. Dilute the sample and use the recommended volume of 10-20 µL per assay.

    Why am I getting a low signal in lysate using a Gaussia/Cypridina/Renilla Luciferase Glow/Luciferase Flash assay? Can you provide some suggestions?

    Here are possible causes and solutions:

    - Non-optimized lysis buffer used: Assay luciferase activity in the media to confirm good expression of luciferase; Use only the provided lysis buffer.
    - Low luciferase expression: Lyse cells in smaller volume of 1X Cell Lysis Buffer; Use a different promoter or growth conditions to improve expression; Increase the integration time on the instrument; Scale up volume of sample and reagent per well.

    I got a low signal in media when I used a Gaussia/Cypridina/Renilla Luciferase Glow/Luciferase Flash assay. What happened?

    Here are possible causes and solutions:

    - Insufficient luciferase accumulation in media: Incubate cells for a longer time.
    - Low luciferase expression: Use less media per well during the experiment;.Use a different promoter or growth conditions to improve expression;.Increase the integration time on the instrument;.Scale-up the volume of sample and reagent per well.
    - Treatment interfered with cellular secretory pathway: Transfect cells with a plasmid for constitutive expression of Luciferase (i.e., with pTK or pCMV promoter); Determine if luciferase actively expresses in media without treatment. Add treatment; Determine if there is a corresponding drop in luciferase activity from the constitutively expressed plasmid.

    I am getting no signal with my Gaussia/Cypridina/Renilla Luciferase Glow/Luciferase Flash assay. Why is this?

    Here are possible causes and solutions:

    - Low transfection efficiency: Optimize transfection conditions using a visual transfection control (e.g., a plasmid over-expressing a fluorescent protein); Verify plasmid DNA quality - use only transfection grade DNA; Use actively dividing, low passage cells; Use a different cell type.
    - No or low promoter activity: Use conditions known for promoter activation; Incubate cells for a longer time; Change growth conditions to improve expression; Use a different promoter.
    - Substrate auto-oxidized: Protect substrate from light and air; Maintain 100X Coelenterazine at -80 degrees C; maintain 100X Vargulin at -80 degrees C; Prepare new Coelenterazine Working Solution if used longer than 8 hours; Prepare new Vargulin Working Solution if used longer than 2 hours

    I prepared my Luciferase Glow Assay Working Solution four hours ago, can I still use it?

    All Luciferase Glow Assay Working Solutions should be stored protected from light.

  • Renilla Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 8 hours at room temperature.
  • Gaussia Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 4 hours at room temperature. 
  • Cypridina Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for up to 2 hours at room temperature.
  • Firefly Luciferase Glow Assay Working Solution must be stored at room temperature (20-25°C) before use and is stable for 4 hours at room temperature. For long term use, store at -20 degrees C for up to 2 months.

  • Does serum concentration have an inhibitory effect on the luciferase assay?

    For Fetal Bovine Serum (FBS), there is no difference between 10% and 5% FBS. It is worthwhile to note that the type of serum does affect the luciferase assay. We tested the effect of 6 different types of sera (FBS, heat-inactivated FBS, dialyzed FBS, charcoal-treated FBS, donor adult bovine serum, and donor equine serum) on secreted luciferases, e.g. Gaussia, Gaussia-Dura, and Cypridina Luciferases. No noticeable difference was observed among different types of sera except for donor adult bovine serum. Unknown factors in donor adult bovine serum caused up to 35% inhibition in secreted luciferase activity. Therefore, we don’t recommend using donor adult bovine serum for Gaussia, Gaussia-Dura, and Cypridina Luciferases.

    I used a white plate to read the luciferase assay and have very high background readings. Should I use a black plate?

    The problems with white plates are related to cross-talk blocking, surface reflectivity and tendency to phosphorescence. We do not recommend using white plates because they give much higher background even if adapted to darkness for 10 minutes. The white plastic does not block the light completely, and some portion of the signal will go through the plastic and will be seen when you measure adjacent wells. The typical situation is if you change from black to white plates, where the background is about doubled and the signal is increased about 10 times. Black plates are recommend for best signal to noise ratio even though the RLU values will be lower. 

    What is the benefit of using the TurboLuc Luciferase One-Step Glow Assay Kit over the Luciferase Glow Assay kits?

    TurboLuc Luciferase One-Step Glow Assay Kit measures activity in mammalian cells with a single reagent-addition step, making it ideal for high-throughput screening (HTS) applications. Several features of the TurboLuc Luciferase One-Step Glow Assay Kit make it suitable for luminometers without injectors and for HTS applications. The convenient, one-step, homogenous protocol minimizes handling steps to support the use of automation. Simply combine the supplied substrate solution and assay buffer to make a single working solution, and then add it to microplate containing the transfected, treated cells. The stable signal output (glow enzyme kinetics) of this system provides flexibility with regard to lead time before read. When used with Thermo Scientific TurboLuc 16 Luciferase vectors, the TurboLuc Luciferase One-Step Glow Assay Kit provides a highly sensitive bioluminescent reporter assay system for the detection of promoter or pathway activity.

    What is the stability of the secreted luciferase?

    Gaussia, Gaussia-Dura, and Cypridina Luciferase are stable in the culture media for greater than 24 hours. Generally, after transfection of CMV driven constructs, we see ever increasing secreted signal between 24 and 72 hours.

    How quickly is luciferase secreted?

    The earliest detectable signal will depend on the strength of the promoter. In general, with a strong promoter (such as CMV), significant signal over background can be seen in as little as 20 minutes. However, with a weaker inducible promoter, significant signal over background may take 1-2 hours after induction.

    What instrument filters are required to read the luciferase assays?

    We always recommend the luciferases to be read without filters for single luciferase assays. Use of filters reduces the amount of signal captured that may lead to decrease in sensitivity. Filters are required only for dual-spectral assays.

    Which luciferase assays can be used to specifically test media as a sample type?

    Gaussia and Cypridina Luciferase assays can be tested on both media and cell lysate sample types. Renilla and Firefly Luciferase assays can only be used on cell lysate as a sample type. 

    Which luciferase assays do you offer to screen cell lysates and media?

    We offer the following luciferase assays (each available in Glow and Flash formats) that provide a highly sensitive assay for transcriptional activity of regulatory elements in mammalian cell culture media and whole cell lysate:

    - Thermo Scientific Pierce Gaussia Luciferase assays provide a highly sensitive system for detecting intracellular and secreted luciferase activity from promoter or pathway activation in mammalian cell culture experiments. Gaussia luciferase has greater protein stability and signal brightness than native Firefly or Renilla luciferase. The bioluminescent signal produced by Gaussia luciferase results from the oxidation of coelenterazine. This reaction does not require adenosine triphosphate (ATP) or other cofactors. The light output correlates with the amount of Gaussia protein expressed, which is proportional to the activity of the promoter for Gaussia expression.
    Note: Gaussia luciferase is a ~22 kDa protein.

    - Thermo Scientific Pierce Renilla Luciferase assays provide a highly sensitive system for detecting intracellular luciferase activity from promoter or pathway activation in mammalian cell culture experiments. The bioluminescent signal produced by green Renilla luciferase results from the oxidation of coelenterazine. This reaction does not require adenosine triphosphate (ATP) or other cofactors. The light output correlates with the amount of green Renilla protein expressed, which is proportional to the activity of the promoter for green Renilla expression.
    Note: Unlike Gaussia and Cypridina, Green Renilla Luciferase can only be assayed in cell lysates as it is not secreted into the cell culture media. Green Renilla luciferase is a ~36 kDa protein.

    - Thermo Scientific Pierce Cypridina Luciferase assays provide a highly sensitive system for detecting intracellular and secreted luciferase activity from promoter or pathway activation in mammalian cell culture experiments. The Cypridina luciferase protein is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. Cypridina luciferase has greater protein stability and signal brightness than native firefly or Renilla luciferase. The bioluminescent signal produced by Cypridina luciferase results from the oxidation of vargulin. This reaction does not require adenosine triphosphate (ATP) or other cofactors. The light output correlates with the amount of Cypridina protein expressed, which is proportional to the activity of the promoter for Cypridina expression.
    Note: Cypridina luciferase is a 61kDa protein.

    - Thermo Scientific Pierce Firefly Luciferase Assays provide a highly sensitive system for detecting intracellular luciferase activity from promoter or pathway activation in mammalian cell culture experiments. The bioluminescent signal produced by firefly luciferase results from the oxidation of D-Luciferin. The light output correlates with the amount of firefly protein expressed, which is proportional to the activity of the promoter for firefly expression.
    Note: Unlike Gaussia and Cypridina, Firefly Luciferase can only be assayed in cell lysates as it is not secreted into the cell culture media.
    Note: Firefly luciferase is a ~60 kDa protein produced in nature by several species of the Lampyridae family of beetles which includes the genera Photinus and Luciola.

    Do you still offer quantitative IP (qIP) Luciferase assay kits?

    Our qIP Luciferase assay kits have been discontinued but we do carry epitope-tagged (HA or c-Myc) and Tluc (TurboLuc luciferase)-tagged mammalian expression vectors, and qIP protein interaction assay reagents for these assay kits.


    compliance-icons compliance-icons compliance-icons
    Product Identifier
    • Renilla Luciferase Glow Assay Kit
    Signal Word
    • Danger
    Hazard Category
    • Acute toxicity Category 3
    • Flammable liquid Category 2
    • Specific target organ toxicity after single exposure Category 1
    Hazard Statement
    • H225-Highly flammable liquid and vapour.
    • H301-Toxic if swallowed.
    • H311-Toxic in contact with skin.
    • H331-Toxic if inhaled.
    • H370-Causes damage to organs.
    Precautionary Statement
    • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P264-Wash thoroughly after handling.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
    • P405-Store locked up.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contain: Methanol

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.