missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RIPA Lysis and Extraction Buffer

Product Code. 10230544 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10230544 250 mL 250mL
10017003 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10230544 Supplier Thermo Scientific™ Supplier No. 89901
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lyse cultured mammalian cells with this high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent.

Thermo Scientific RIPA Lysis and Extraction Buffer is a high-quality, ready-to-use and fully disclosed formulation of a popular cell lysis reagent for cultured mammalian cells.

Features of RIPA Buffer:

  • Convenient—ready-to-use solution; no need to assemble and prepared components yourself
  • Flexible—compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification
  • Versatile—enables extraction of cytoplasmic, membrane and nuclear proteins
  • Disclosed formulation—contains no proprietary components, providing users with complete control and knowledge of possible compatibility issues

This RIPA buffer effectively lyses and extracts protein from cultured mammalian cells, including plated cells and pelleted suspension cells. The popular reagent enables the extraction of membrane, nuclear and cytoplasmic proteins and is compatible with many applications, including reporter assays, the Thermo Scientific BCA Protein Assay (Cat. No. 23225), immunoassays and protein purification. Inhibitors such as Thermo Scientific Halt Protease Inhibitor Cocktail (Cat. No. 78420) and Halt Phosphatase Inhibitor Cocktail (Cat. No. 78420) are also compatible with this RIPA buffer formulation and can be added before use to prevent proteolysis and maintain protein phosphorylation.

RIPA buffer derives its name from the original application for which it was developed: the radio-immunoprecipitation assay. While this isotopic assay method is rarely performed in laboratories today, the acronym for this lysis buffer formulation has endured in common use. RIPA cell lysis reagent is highly effective for protein extraction from a variety of cell types because it contains three non-ionic and ionic detergents. One disadvantage of this detergent formulation is its relative incompatibility with certain downstream applications compared to other lysis reagents.

Related Products

  • Pierce™ IP Lysis Buffer (Cat. No. 87787)
  • M-PER™ Mammalian Protein Extraction Reagent (Cat. No. 78501)
TRUSTED_SUSTAINABILITY

Specifications

Quantity 250 mL
Format Liquid
Product Type Extraction Buffer
Volume (Metric) 250 mL
Content And Storage Upon receipt store at 4°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.