missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ SATA (N-succinimidyl S-acetylthioacetate)

Product Code. 10750805 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10750805 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10750805 Supplier Thermo Scientific™ Supplier No. 26102
Spotlight Offers
Use promo code 28037
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Short (2.8A), amine-reactive (NHS ester) sulfhydryl-addition reagent; creates stable (protected yet exposable) -SH group for protein crosslinking strategies.

Thermo Scientific™ Pierce SATA is a short-chain (2.8 Å spacer arm) reagent for covalent modification of primary amines and addition of a stable yet exposable sulfhydryl group to facilitate heterobifunctional crosslinking strategies.

Highlights:

  • Reacts with primary amines to add protected sulfhydryls
  • An easy deprotection step generates a free sulfhydryl group
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine
Content And Storage Upon receipt store desiccated at -20°C. Product is shipped at ambient temperature.
Label or Dye Sulfhydryl Group
Label Type Conjugating Groups
Product Line Pierce
Crosslinker Type Heterobifunctional
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer Short (<10 Å)
Form Powder
Reagent Type SATA
Quantity 50 mg
Solubility DMF, DMSO
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.