missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Small Molecule System Suitability Standard

Product Code. 17767453 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 injections, 0.25 mL
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17767453

Brand: Thermo Scientific™ A51740

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Thermo Scientific™ Pierce™ Small Molecule System Suitability Standard provides a pre-formulated mixture of nine small-molecule standards to assess system performance in both positive and negative ionization modes for TSQ Triple Quadrupole and Orbitrap Exploris™ mass spectrometers.

  • A pre-formulated mixture of nine small-molecule standards to assess system performance in both positive and negative ionization modes for TSQ Triple Quadrupole and Orbitrap Exploris mass spectrometers
  • Comprehensive mixture of nine small-molecule compounds (mass range: 76 to 1279)
  • Designed to assess instrument performance in both positive and negative ionization modes
  • Highly validated, high-quality, rigorously-tested formulation with lot-specific certificates of analysis
  • Convenient, pre-formulated ready-to-use liquid format
  • An excellent QC assay tool for quality assessment of LC and MS instrument status
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Quantity 10 injections, 0.25 mL
Grade MS Grade
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.