missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium Deoxycholate Detergent

Product Code. 11864141 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11864141 5 g 5g
11874141 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11864141 Supplier Thermo Scientific™ Supplier No. 89904
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Water-soluble, ionic detergent used in cell lysis buffers (e.g., RIPA buffer) to disrupt protein interactions for liposome and membrane protein isolation.

Thermo Scientific™ Sodium Deoxycholate is an ionic detergent that is especially useful for disrupting and dissociating protein interactions.

Features of Sodium Deoxycholate:

  • Popular anionic, bile-acid detergent for many laboratory uses
  • Effective in disrupting and dissociating many types of protein interaction
  • Can be removed from solution by dialysis
  • Useful for elution or regeneration of certain kinds of affinity columns
  • High-purity compound with low UV absorptivity

Properties of Sodium Deoxycholate:

  • Alternative Names: Sodium deoxycholic acid; deoxycholate, sodium salt
  • Chemical Name: 3, 12-α-Dihydroxy-5β-cholan-24-oic acid, monosodium salt
  • Molecular Weight: 414.6 g
  • Detergent Class: Ionic (anionic)
  • Aggregation Number: 5 (average)
  • Micelle Molecular Weight: 2000 g (average)
  • Critical Micelle Concentration (CMC): 2 to 6 mM (0.083 to 0.249%, w/v)
  • Cloud Point: Unknown
  • Dialyzable: Yes

Specifications for Sodium Deoxycholate (Part No. 89904, 89905):

  • Formula: C24H39O4Na
  • Molecular Weight: 414.6 g
  • Purity (by HPLC): ≥98%
  • Absorbance (1% Detergent Solution): 340 nm <0.02; 280 nm <0.04; 260 nm <0.06
  • pH (1% Solution): 5 to 9
  • Solubility (in water at 20°C): ≥5%

Sodium deoxycholate (deoxycholic acid) is a water-soluble, bile-acid, ionic detergent commonly used in protein methods. It is most frequently used as a component of cell lysis buffers (e.g., RIPA buffer), but also has been used for liposome preparation, isolation of membrane proteins and lipids, preventing non-specific binding in affinity chromatography and a cell culture media supplement.

The effectiveness of a detergent in any application depend upon the detergent's concentration. Too much or too little detergent can often have a deleterious effect. It is recommended that you examine a variety of detergent concentrations in your application. At concentrations above 2 mM, cholate will form micelles having MW ∼2000. The small micelle size allows easy removal by dialysis or gel filtration when needed. Note: removal of a detergent from a protein solution may result in protein precipitation and/or aggregation.

Sodium deoxycholate is the detergent recommended for stripping endotoxin (Lipopolysaccharide or LPS) from immobilized Polymyxin B columns. This is the recommended product for use with the Thermo Scientific Detoxi-Gel Endotoxin Removing Gel.

Related Products

Surfact-Amps™ Detergent Sampler (Cat. No. 28340)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Detergent
Content And Storage Store at -20°C
Form Powder
Reagent Type Detergent (Pure)
Quantity 5 g
compliance-icons
Product Identifier
  • Sodium Deoxycholate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash hands thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium 3-droxy-5-oate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.