missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SPB (succinimidyl-[4-(psoralen-8-yloxy)]-butyrate)

Product Code. 10627534 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
10627534 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 10627534

Brand: Thermo Scientific™ 23013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Crosslink amines to DNA with this crosslinker composed of NHS-ester and psoralen reactive groups; conjugates to DNA via light-activated intercalation next to pyrimidine bases.

Thermo Scientific™ Pierce SPB is an NHS-ester and psoralen crosslinker that effectively conjugates primary amines to DNA via light-activated intercalation next to pyrimidine bases.

Highlights:

  • Melting point: 177-178°C
  • Reactive groups: NHS ester and psoralen (Amax= 282nm)
  • Reactive toward: amino groups pyrimidines in DNA or RNA
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Nonselective
Cleavable No
Description SPB
Molecular Weight (g/mol) 385.32
PEGylated No
Spacer Arm Length 8.6 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Not determined
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Psoralen, NHS Ester
Spacer Short (<10 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.