Learn More
Description
Pierce™ Spin Tip Adaptors
Spin tip adaptors are designed to securely hold spin tips in standard microcentrifuge tubes during centrifugation. The adaptors help to ensure proper positioning of the tip, providing consistent flow-through and efficient sample processing across activation, equilibration, washing, and elution steps.
Compatible with a range of spin tip chemistries—including C18, SDB, SAX, SCX, and MCX—these adaptors support flexible sample preparation workflows for proteomics and other applications.
Features of spin tip adaptors
Reliable – secure fit for spin tips during centrifugation
Compatible – designed for standard microcentrifuge tubes
Efficient – Enables consistent flow-through and recovery
Spin tip adaptors allow for simple integration into existing workflows.
Specifications
Specifications
| For Use With (Equipment) | Microcentrifuge, Spin Tips, Microcentrifuge Tube |
| Product Type | Spin Tip Adaptor |
| Product Line | Pierce |
| Content And Storage | Adaptors |
| Shipping Condition | Room Temperature |
| Starting Material | Peptides |
| Detection Method | Mass Spectrometry |
| Format | Insert (Refill) |
| Quantity | 25 Pk. |
| For Use With (Application) | Peptide Desalting |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.