missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin, Maleimide-Activated

Product Code. 10321204 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10321204 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321204 Supplier Thermo Scientific™ Supplier No. 21102
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Perform Western blot, ELISA or other biotin-based detection with purified and HRP-, AP-, FITC-, TRITC-, PE- or APC-conjugated or maleimide-/hydrazide-activated streptavidin.

Thermo Scientific™ Pierce Streptavidin and Conjugates include purified recombinant streptavidin biotin-binding protein, plus standard SA-HRP, SA-AP and fluorescent conjugates for biotin-based assays.

These purified and conjugated forms of recombinant streptavidin protein enable detection of biotinylated antibodies and other probes in a variety of standard assay methods, including Western blotting, ELISA, immunohistochemistry and fluorescence imaging. Purified streptavidin and the streptavidin-HRP conjugates (peroxidase-conjugated streptavidin) are supplied lyophilized in phosphate-based buffers for immediate reconstitution with water. Alkaline Phosphatase conjugates are supplied as a liquid in a Tris-based buffer.

Highlights:

  • Recombinant streptavidin – high-quality protein, 53kDa, pI equals 6.8 to 7.5

  • Pure protein and enzyme conjugates – choose pure, unconjugated protein for biotin blocking and controls; enzyme conjugates for substrate-based detection

  • Fluorophore conjugates – standard fluorescent forms (fluorescein, rhodamine, etc.) for traditional immunofluorescence techniques

  • Activated preparations – choose a maleimide-activated or hydrazide-activated form to prepare a custom streptavidin conjugate with sulfhydryl- or carbohydrate-containing molecule

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Streptavidin
Description Pierce Streptavidin, Maleimide Activated
Formulation Lyophilized powder from protein in buffered saline solution
Quantity 1 mg
Content And Storage Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature.
Chemical Reactivity thiol
Reactive Moiety Maleimide
Solubility Soluble in Water
Form Solid
Product Line Pierce
Product Type Streptavidin Conjugate
Label or Dye Streptavidin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.