missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-GMBS (N-γ-maleimidobutyryl-oxysulfosuccinimide ester)

Product Code. 11881285 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11881285 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11881285 Supplier Thermo Scientific™ Supplier No. 22324
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Crosslink amines to sulfhydryls with this short, water-soluble crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of an aliphatic, 6-atom (7.3A) spacer arm.

Thermo Scientific™ Pierce Sulfo-GMBS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a short spacer arm (7.3 Ås).

Highlights:

  • Has a low potential for eliciting crosslinker initiated immune response
  • GMBS and Sulfo-GMBS are less immunogenic than SMCC
  • Soluble in water and aqueous buffer systems
  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Description Sulfo-GMBS
Molecular Weight (g/mol) 382.28
PEGylated No
Spacer Arm Length 7.3 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short (<10 Å)
Form Powder
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.