missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-LC-SPDP (sulfosuccinimidyl 6-(3'-(2-pyridyldithio)propionamido)hexanoate)

Product Code. 11801255 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11801255 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11801255 Supplier Thermo Scientific™ Supplier No. 21650
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Crosslink amines to sulfhydryls with this water-soluble sulfo-NHS-ester and pyridyldithiol groups on 10-atom spacer arms to form cleavable disulfide bonds with cysteines.

Thermo Scientific™ Pierce Sulfo-LC-SPDP is a water-soluble, long-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteines.

Highlights:

  • Forms cleavable disulfide bonds with sulfhydryls
  • Releases a detectable byproduct after reacting with free sulfhydryl groups; by measuring the release of pyridine-2-thione at 343nm, the reaction progression can be monitored
  • Reactive groups: sulfo-NHS ester and pyridyldisulfide
  • Reactive toward: amino and sulfhydryl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable By Thiols
Description Sulfo-LC-SPDP
Molecular Weight (g/mol) 527.57
PEGylated No
Spacer Arm Length 15.7 Å
Content And Storage Upon receipt store at -20°C protected from moisture.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Pyridyldisulfide, Sulfo-NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.