missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Sulfo-NHS-Acetate

Product Code. 10618494 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10618494 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10618494 Supplier Thermo Scientific™ Supplier No. 26777
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amine-reactive (NHS ester) compound to permanently block primary amines (e.g., protein lysine side chains) to prevent interactions or later conjugation.

Thermo Scientific™ Pierce Sulfo-NHS-Acetate is a small compound that forms stable, covalent amide bonds with primary amines (e.g., lysine side chains), enabling them to be permanently blocked to prevent interactions or later conjugation.

Highlights:

  • Reacts mainly with primary amines (lysine ε amino groups)
  • Irreversible reaction is accompanied by the release of N-hydroxysulfosuccinimide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine
Content And Storage Upon receipt store desiccated at -20° C. Product is shipped at ambient temperature.
Label or Dye Unreactive Group
Label Type Blocking Groups
Product Line Pierce
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer Short (<10 Å)
Form Powder
Quantity 100 mg
Solubility DMSO, Water, DMF
Product Type Sulfo-NHS-Acetate
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.