missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulfo-SMPB (sulfosuccinimidyl 4-(N-maleimidophenyl)butyrate)

Product Code. 11851285 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11851285 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11851285 Supplier Thermo Scientific™ Supplier No. 22317
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Crosslink amines to sulfhydryls with this mid-length, water-soluble crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of an aromatic, 10-atom spacer arm.

Thermo Scientific™ Pierce Sulfo-SMPB is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length aromatic spacer arm (11.6 Ås).

Highlights:

  • Extended chain length limits steric hindrance
  • Water-soluble; membrane-impermeable
  • Reactive groups: sulfo-NHS ester and maleimide
  • Reactive toward: amino and sulfhydryl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Sulfhydryl
Cleavable No
Description Sulfo-SMPB
Molecular Weight (g/mol) 458.37
PEGylated No
Spacer Arm Length 11.6 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.