missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TNBSA Solution (2,4,6-trinitrobenzene sulfonic acid) (5% w/v)

Artikelnummer. 10210314 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
10210314 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10210314 Lieferant Thermo Scientific™ Lieferanten-Nr. 28997

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Label primary amines (peptides or amino acids) for colorimetric detection (Amax 335nm) using this 5% solution of trinitrobenzene sulfonic acid in methanol.

Thermo Scientific™ Pierce TNBSA is a 5% solution of trinitrobenzene sulfonic acid in methanol that reacts with primary amines (peptides or amino acids) to yield a soluble colored product, a property useful for various assay methods.

Highlights:

  • Couples with primary amines, sulfhydryls and hydrazides in aqueous solution at pH 8, without undesirable side reactions
  • Excellent for solution or solid phase analysis
  • Suitable for qualitative and quantitative estimation of biomolecules
  • λmax = 335nm
  • Reacts readily with primary amino groups of amino acids in aqueous format at pH 8 to form yellow adducts
  • No colored derivatives are formed with secondary amino acids proline and hydroxyproline
  • Colored derivatives are monitored at 345nm and have extinction coefficients in range of 1-1.5 x 104
TRUSTED_SUSTAINABILITY

Spezifikation

Product Type TNBSA Solution
Chemical Reactivity Amine
Content And Storage Storage: Upon receipt store at -20°C. Product is shipped at ambient temperature.
Label or Dye Other Label or Dye
Detection Method Colorimetric
Quantity 100 mL
Form Liquid

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.