missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TNBSA Solution (2,4,6-trinitrobenzene sulfonic acid) (5% w/v)

Product Code. 13427086 Shop All Thermo Scientific Products
Change view
Click to view available options
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
13427086 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13427086 Supplier Thermo Scientific™ Supplier No. TS28997
Quantity Price
1 370.26 € / 100mL
6 302.64 € / 100mL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Label primary amines (peptides or amino acids) for colorimetric detection (Amax 335nm) using this 5% solution of trinitrobenzene sulfonic acid in methanol.

Thermo Scientific™ Pierce TNBSA is a 5% solution of trinitrobenzene sulfonic acid in methanol that reacts with primary amines (peptides or amino acids) to yield a soluble colored product, a property useful for various assay methods.

Highlights:

  • Couples with primary amines, sulfhydryls and hydrazides in aqueous solution at pH 8, without undesirable side reactions
  • Excellent for solution or solid phase analysis
  • Suitable for qualitative and quantitative estimation of biomolecules
  • λmax = 335nm
  • Reacts readily with primary amino groups of amino acids in aqueous format at pH 8 to form yellow adducts
  • No colored derivatives are formed with secondary amino acids proline and hydroxyproline
  • Colored derivatives are monitored at 345nm and have extinction coefficients in range of 1-1.5 x 104
TRUSTED_SUSTAINABILITY

Specifications

Product Type TNBSA Solution
Chemical Reactivity Amine
Content And Storage Storage: Upon receipt store at -20°C. Product is shipped at ambient temperature.
Label or Dye Other Label or Dye
Detection Method Colorimetric
Quantity 100 mL
Form Liquid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • TNBSA Solution (2,4,6-trinitrobenzene sulphonic acid)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use dry sand, dry chemical, or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2,4,6-trinitrobenzenesulphonic acid, methyl alcohol

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.