missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Universal Nuclease for Cell Lysis

Product Code. 12156490 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 kU
25 kU
5 kU
Unit Size:
100ku
25ku
5ku
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12156490 25 kU 25ku
12146490 5 kU 5ku
12391963 100 kU 100ku
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12156490 Supplier Thermo Scientific™ Supplier No. 88701
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for a wide variety of applications where complete digestion of nucleic acids is needed and can be used in conjunction with cell lysis buffers.

Thermo Scientific Pierce Universal Nuclease for Cell Lysis is ideal for a wide variety of applications where complete digestion of nucleic acids is needed when preparing cell lysates.

Features of Universal Nuclease for Cell Lysis:

  • Broad spectrum—degrades all forms of DNA and RNA
  • Highest-quality enzyme—nuclease is ≥99% pure, as tested by SDS-PAGE
  • Robust activity—100-fold greater specific activity than DNase I
  • Versatile—can be used with a wide variety of cell lysis reagents

Pierce Universal Nuclease for Cell Lysis is a genetically engineered endonuclease from Serratia marcescens. The enzyme is produced and purified from E. coli and consists of two identical 30-kDa subunits with two critical disulfide bonds. This indiscriminate endonuclease degrades single-stranded, double-stranded, linear and circular DNA and RNA and is effective over a wide range of temperatures and pH. This enzyme has high specific activity (100-fold greater than DNase I) and increased thermal stability compared to other nucleases. Pierce Universal Nuclease is ≥99 pure enzyme, is free of any measurable protease activity and is supplied at 250 U/μL. Pierce Universal Nuclease for Cell Lysis is identical in performance to Benzonase™ Nuclease (EMD Merck).

Applications:

  • Use with B-PER, Y-PER or other commercial or homebrew cell lysis reagents and/or mechanical disruption to reduce viscosity in protein extracts
  • Remove DNA and RNA from recombinant protein preparations prior to downstream processing

Pierce Universal Nuclease for Cell Lysis is commonly used to reduce the viscosity of bacterial and mammalian protein extracts for downstream application by removing the nucleic acids from protein preparations. The enzyme completely digests nucleic acids to oligonucleotides that are less than 5 bases long. Pierce Universal Nuclease for Cell Lysis helps to improve the separation of the lysate pellet from the supernatant, enhances filtration of the treated lysate, improves chromatography processing time and increases the overall protein yield. The endonuclease has also been shown to improve the compatibility of protein extracts for 2D gel electrophoresis. One unit corresponds to the amount of enzyme required to produce a change of 1.0 in the absorbance at 260 nm of sonicated Herring DNA over 30 minutes at 37°C, as determined using standard nuclease from the Merck™ Serratia marcescens volumetric activity assay.

Related Products

Micrococcal Nuclease Solution (≥ 1 unit/μL) (Cat. No. 88216)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cell Lysis Enzyme
Content And Storage Store in a cool, dry, well-ventilated area, protected from direct sunlight.
Form Liquid
Product Line Pierce
Buffer Lysis Buffer
Reagent Type Enzyme for Cell Lysis
Enzyme Nuclease
Quantity 25 kU

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.