missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Water, LC-MS Grade

Product Code. 11591094 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11591094 1 L 1 L
10763295 1 L 1 L
13484279 4 x 1 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11591094 Supplier Thermo Scientific™ Supplier No. TS51140

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC

Thermo Scientific™ Water, LC/MS Grade is ultapure water for formulation of solvents for HPLC.

An ultrapure, LC/MS-grade preparation with low UV-absorptivity that makes it suitable and reliable for use in HPLC and mass spectrometry applications.

Thermo Scientific Water, LC/MS Grade is specially purified by a proprietary method and treated to ensure lot-to-lot consistency with a low UV absorbance to provide you with most sensitive detection across all wavelengths. Pierce Water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmoshere with Teflon TFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Purity and quality validated with 30 tests for use in LC/MS applications
  • 0.2-micron filtered, packed in solvent-rinsed amber glass bottles
  • Sealed under nitrogen atmosphere to eliminate absorption of unknown atmospheric gases

Product Details:

General properties of water

  • Molecular formula: H2
  • Molecular Weight: 18.01
  • CAS number: 7732-18-5

Water, LC/MS Grade is specifically purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.

TRUSTED_SUSTAINABILITY

Spezifikation

pH 6 to 8
Color Clear, Colorless
Melting Point 32°F
Boiling Point 212°F
Grade LC-MS
Quantity 1 L
Physical Form Liquid
Product Line Pierce
What kind of buffers and solutions are compatible with peptide clean-up and LC-MS?

Most organic solvents or volatile buffers are compatible with LC-MS. Water and acetonitrile with 0.1% formic acid is the most common solvent system for separation of peptide samples for mass spectrometry analysis. Trifluoracetic acid (TFA) is an alternative ion pairing agent for off-line peptide C18 clean-up or LC-UV analysis. We offer the following LC-MS grade reagents: Pierce 0.1% Trifluoroacetic Acid (v/v) in Water (Cat. No. 85172), Pierce Water, LC-MS Grade (Cat. No. 85189), Pierce Acetonitrile (ACN) (Cat. No. 85188), and Pierce Trifluoroacetic (TFA) (Cat. No. 85183).

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.