missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Western Blot Roller

Product Code. 15195204 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15195204 1 unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15195204 Supplier Thermo Scientific™ Supplier No. 84747
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A simple, reusable tool for gently and easily removing trapped air bubbles when assembling transfer stacks in semi-dry or wet-tank transfer cassettes.

The Thermo Scientific™ Western Blot Roller is a simple, reusable tool for gently and easily removing trapped air bubbles when assembling transfer stacks in semi-dry or wet-tank transfer cassettes.

The transfer-stack roller has a stainless steel handle and an inert PTFE roller that is 8.4cm wide. Mini-gel sandwiches can be rolled out with one or two passes, but the tool also works well for transfer stacks with larger gels. The tool is included as an accessory with the Pierce G2 Fast Blotter (Part No. 62288).

Highlights:

  • Stainless steel handle
  • PTFE roller, 8.4cm wide
  • Reusable laboratory tool
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage A 8.4 cm-wide PTFE roller with stainless steel handle
For Use With (Equipment) Pierce Power System
Mode of Transfer Semi-Dry, Wet
Type Western Blot Roller
Quantity 1 unit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.