missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Yeast DNA Extraction Kit

Product Code. 10473985 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
75 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10473985 75 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10473985 Supplier Thermo Scientific™ Supplier No. 78870
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Quickly, efficiently and easily lyse yeast cells to release DNA with Y-PER lysis reagent.

Quickly and easily lyse yeast cells to release DNA for study with Thermo Scientific™ Pierce Yeast DNA Extraction Kit, which utilizes a Thermo Scientific γ-PER Reagent.

The kit provides greater yields than traditional methods of DNA isolation from yeast. Typically, extraction and purification of DNA from yeast are time-consuming and labor-intensive. The yeast proteinaceous cell wall is notoriously difficult to lyse and requires harsh treatments that are time-consuming and can damage the extracted DNA. The Yeast DNA Extraction Kit protocol requires less than one hour to complete, is effective without enzymatic treatment or glass beads, and yields little to no RNA contamination regardless of RNase presence. In studies with Saccharomyces cerevisiae, high yields of genomic and plasmid DNA are consistently obtained. DNA purified using this kit is suitable for PCR amplification, bacterial transformations, restriction digestions and hybridization applications.

Highlights:

  • Eliminates need for glass beads or harsh enzyme treatments
  • Yields DNA that is compatible with polymerase chain reaction (PCR) amplification
  • Rapidly isolates plasmid DNA from S. cerevisiae suitable for transformation of E. coli
  • Protocol is easily scalable for use with single colonies or 500mL cultures

Includes:

Thermo Scientific γ-PER Yeast Protein Extraction Reagent, DNA releasing reagents A and B, and protein removal reagent

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: 50 purifications from 10 mL cultures
• Y-PER Yeast Protein Extraction Reagent, 25 mL
• DNA Releasing Agent A, 20 mL
• DNA Releasing Agent B, 20 mL
• Protein Removal Reagent, 10 mL

Store at 4°C. Precipitates may form; gently warm at 37°C for 1–5 minutes.
Sample Type Yeast
For Use With (Application) PCR, Sequencing, Transformation
High-throughput Compatibility Not High-throughput Compatible
Product Type Yeast DNA Extraction Kit
Quantity 75 mL
Target Genomic DNA
compliance-icons
Product Identifier
  • Yeast DNA Extraction Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: lithium chloride, sodium dodecyl, trometamol, sodium chloride, sodium chloride,disodium hydrogenort

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.