missing translation for 'onlineSavingsMsg'
Learn More

Pifithrin-β hydrobromide, MedChemExpress

Product Code. 16469677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469677 5 mg 5mg
16427148 10 mg 10mg
16403177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16469677 Supplier Medchem Express Supplier No. HY16702A
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pifithrin-β hydrobromide (PFT β hydrobromide) is a potent p53 inhibitor with an IC50 of 23 μM.

Pifithrin-β hydrobromide (PFT β hydrobromide) is a potent p53 inhibitor with an IC50 of 23 μM.

Specifications

Chemical Name or Material Pifithrin-β hydrobromide
CAS 511296-88-1
Color White
Purity Grade Notes Research
Molecular Formula C16H17BrN2S
Quantity 5 mg
Synonym PFT β hydrobromideCyclic Pifithrin-α hydrobromide
Solubility Information DMSO : 10 mg/mL (28.63 mM; Need ultrasonic)
SMILES CC1=CC=C(C2=CN3C(SC4=C3CCCC4)=N2)C=C1.Br
Molecular Weight (g/mol) 349.29
Formula Weight 349.29
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.