missing translation for 'onlineSavingsMsg'
Learn More

Pigment Green 7 (Technical Grade), TRC

Product Code. 30425238
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30425238 500 mg
30424157 5 g
30422590 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425238 Supplier Toronto Research Chemicals Supplier No. TRCP437833500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1328-53-6
Molecular Formula C32Cl16CuN8
Molecular Weight (g/mol) 1127.19
SMILES ClC1=C(Cl)C(Cl)=C(Cl)C2=C3/N=C\4C5=C(Cl)C(Cl)=C(Cl)C(Cl)=C5C(=N4)N=C6C7=C(Cl)C(Cl)=C(Cl)C(Cl)=C7C8=NC(=N9)C%10=C(Cl)C(Cl)=C(Cl)C(Cl)=C%10C9=NC(=C12)N3[Cu]N68

Specifications

Quantity 500 mg
Synonym C.I. Pigment Green 7, 10066064 FDA Green, AF Green E 1, Accosperse Cyan Green G, Acnalin Supra Green FG, Acron Green 4G, Aquafine Green E 1, Brilliant Green phthalocyanine, C.I. 74260, C.I. Pigment Green 42, Calcotone Green G, Cartaren C-VB, Cartaren Green C-VB, Ceres Green 3B, Chromaflo 888-5511, Chromatex Green G, Chromofine Cyanine Green 2GN, Chromofine Green 2GN, Chromofine Green 2GO, Chromofine Green 5301, Clear Green L, Colanyl Green GG, Colortex Green P 1011, Copper Phthalocyanine Green, Cosmenyl Green GG, Cromophtal Green GF, Cromophtal Green GFN, Cromophtal Green GFN-P, Cyan Green 15-3100, Cyanine Green 2G550D, Cyanine Green 2GN, Cyanine Green 2GO, Cyanine Green 5310, Cyanine Green GP, Cyanine Green NB, Cyanine Green PBN 1, Cyanine Green T, Cyanine Green Toner, Cyanine Green YL Super, Cyanine Green YS 07, DG 007-448, Dainichi Cyanine Green 537, Dainichi Cyanine Green FG, Dainichi Cyanine Green FGH, Daltolite Fast Green GN, Dispers Green 87-3007, Duratint Green 1001, Dymic MBR 510, EB Green B 4701, EM Color Green B, Eupolen PE Green K 87-3001, Fastogen Green 5005, Fastogen Green 5710, Fastogen Green 5716, Fastogen Green B, Fastogen Green S, Fastogen Green SF, Fastogen Green SO, Fastolux Green, Fenalac Green G, Fenalac Green G Disp, Flexiverse Green GFD 0701, Flexobrite C 7, Fuji AS Green, Fuji SP Green 7051, Full Green S, G 5319, Granada Green Lake GL, Graphtol Green 2GLS, Graphtol Green 5869-3, Green 19089, Green GNX-D, Green GNX-TS, Green YS 2A, Heliogen Green 8680, Heliogen Green 8681K, Heliogen Green 8682T, Heliogen Green 8730, Heliogen Green 8730K, Heliogen Green A, Heliogen Green D 8725, Heliogen Green D 8730, Heliogen Green G, Heliogen Green GA, Heliogen Green GN, Heliogen Green GNA, Heliogen Green GTA, Heliogen Green GV, Heliogen Green GWS, Heliogen Green K 8730, Heliogen Green L 8605, Heliogen Green L 8730, Heliogen Green L 8731, Heliogen K 8730, Heuco Green 600703K, Heuco Green 600736, Hostaperm Green GG, Hostaperm Green GG 01, Hostaperm Green GNX, Hostaperm Green GNX-D, Hostaperm Green GNX-L, Hostaperm Green GNX-TS, Hostatint Green GG 30, Irgalite Fast Brilliant Green 3GL, Irgalite Fast Brilliant Green GL, Irgalite Green GFNP, Irgalite Green GLN, Irgalite Green GLNP, Irgalite Green GLPO, Irgazin Green 2180, K 8730, KET Green 201, Klondike Yellow X 2261, Liofast Green B 237, Lionol Green 8120, Lionol Green 8200, Lionol Green 8930, Lionol Green B 201, Lionol Green GLS, Lionol Green POG-T, Lionol Green S, Lionol Green Y 101, Lionol Green Y 102, Lionol Green YS 07, Lionol Green YS 10, Lionol Green YS 2A, Lionol Green YS 7, Luconyl Green 7830, Luconyl Green 8730, Lutetia Fast Emerald J, Microlen Green GFN-UA, Microlite Green GK, Microlith Green G-A, Microlith Green G-FP, Microlith Green G-K, Microlith Green G-KP, Microlith Green G-T, Microlith Green GN, Monarch Green WD, Monastral Fast Green 2GWD, Monastral Fast Green BGNA, Monastral Fast Green G, Monastral Fast Green GD, Monastral Fast Green GF, Monastral Fast Green GFNP, Monastral Fast Green GN, Monastral Fast Green GNA, Monastral Fast Green GTP, Monastral Fast Green GV, Monastral Fast Green GWD, Monastral Fast Green GX, Monastral Fast Green GXB, Monastral Fast Green GYH, Monastral Fast Green LGNA, Monastral Green B, Monastral Green B Pigment, Monastral Green G, Monastral Green GBX-C, Monastral Green GFN, Monastral Green GH, Monastral Green GN, Monastral Green GN-C, Monastral Green GNE-HD, Monastral Green GNX-C, Monolite Fast Green GVSA, Monolite Green 600734, Monolite Green GLX, Monolite Green GN, Non-Flocculating Green G 25, Novotex Green GNX, O 271, Opaline Green G 1, P 6100, P 6100 (pigment), PG 7, PV Fast Green GG, PV Fast Green GNX, PV-Fast Green G, Permanent Green Toner GT-376, Phthalo Green Pigment 264-3100, Phthalo green, Phthalocyanine Brilliant Green, Phthalocyanine Green 2724, Phthalocyanine Green 2GN, Phthalocyanine Green 2GNL, Phthalocyanine Green 5329, Phthalocyanine Green B 308, Phthalocyanine Green G, Phthalocyanine Green LX, ...
InChI Formula InChI=1S/C32Cl16N8.Cu/c33-9-1-2(10(34)18(42)17(9)41)26-49-25(1)53-27-3-4(12(36)20(44)19(43)11(3)35)29(50-27)55-31-7-8(16(40)24(48)23(47)15(7)39)32(52-31)56-30-6-5(28(51-30)54-26)13(37)21(45)22(46)14(6)38;/q-2;+2
Formula Weight 1118.46
Chemical Name or Material Pigment Green 7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.