missing translation for 'onlineSavingsMsg'
Learn More

Pigment Red 170, TRC

Product Code. 30411041
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411041 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411041 Supplier Toronto Research Chemicals Supplier No. TRCP4378051G
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pigment Red 170
CAS 2786-76-7
Molecular Formula C26 H22 N4 O4
Quantity 1 g
Synonym 2-Naphthalenecarboxamide','4-[[4-(aminocarbonyl)phenyl]azo]-N-(2-ethoxyphenyl)-3-hydroxy- (9CI), 2-Naphtho-o-phenetidide','4-[(p-carbamoylphenyl)azo]-3-hydroxy- (7CI,8CI), 4-[2-[4-(Aminocarbonyl)phenyl]diazenyl]-N-(2-ethoxyphenyl)-3-hydroxy-2-naphthalenecarboxamide, 4708 Permanent Red F 5RK, Brilliant Carmine 7010, C.I. 12475, C.I. PR 170, C.I. Pigment Red 170, Eljon Red NRO, Euderm Red, F 2RK, F 3RK70, Fuji Fast Red 2200, Graphtol Red F 3RK70, Graphtol Red F3RK 70-CN09, Graphtol Red NFB, Hostaperm Red F 5RK, JHR 1703L, KP Plast P-Red 3RH, L 1/8 Red F 3RK70, Lysopac Red 7030C, Naphthol AS Red, Naphthol Red 170, Naphthol Red 3172, Naphthol Red 5R, Naphtol Red B 7032C, Novoperm Red F 2RK70, Novoperm Red F 2RK70N, Novoperm Red F 3RK, Novoperm Red F 3RK70, Novoperm Red F 5RK, Novoperm Red F 6RK, PR 170, Permanent Red BH 5RK, Permanent Red F 3RK, Permanent Red F 3RK70, Permanent Red F 5RK, Permanent Red P-FK, Pigment Red 170, Red F 3RK70, Red F 5RK, Sanyo Brilliant Carmine 7009, Seikafast Red 3820, Seikafast Red 3891, Suimei Fast Red ER, Sumitone Fast Red BRS, Sunbrite Red 170
InChI Formula InChI=1S/C26H22N4O4/c1-2-34-22-10-6-5-9-21(22)28-26(33)20-15-17-7-3-4-8-19(17)23(24(20)31)30-29-18-13-11-16(12-14-18)25(27)32/h3-15,31H,2H2,1H3,(H2,27,32)(H,28,33)
SMILES CCOc1ccccc1NC(=O)c2cc3ccccc3c(N=Nc4ccc(cc4)C(=O)N)c2O
IUPAC Name 4-[(E)-(4-carbamoylphenyl)diazenyl]-N-(2-ethoxyphenyl)-3-hydroxynaphthalene-2-carboxamide
Molecular Weight (g/mol) 454.477
Formula Weight 454.164
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.