missing translation for 'onlineSavingsMsg'
Learn More

Pilocarpic Acid Sodium Salt, TRC

Product Code. 30134136
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134136 50 mg 50mg
30040520 10 mg 10mg
30012295 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134136 Supplier Toronto Research Chemicals Supplier No. TRCP44145050MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92598-79-3
Molecular Formula C11 H17 N2 O3 . Na
Molecular Weight (g/mol) 248.25
Synonym (2S,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt, Pilocarpine Hydrochloride Imp. B (EP) as Sodium Salt, Pilocarpic Acid Sodium Salt, 1H-Imidazole-5-butanoic acid, α-ethyl-β-(hydroxymethyl)-1-methyl-, monosodium salt, [S-(R*,S*)]- (9CI), Sodium pilocarpate
IUPAC Name sodium;(2S,3R)-2-ethyl-3-(hydroxymethyl)-4-(3-methylimidazol-4-yl)butanoate
SMILES [Na+].CC[C@@H]([C@H](CO)Cc1cncn1C)C(=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H18N2O3.Na/c1-3-10(11(15)16)8(6-14)4-9-5-12-7-13(9)2;/h5,7-8,10,14H,3-4,6H2,1-2H3,(H,15,16);/q;+1/p-1/t8-,10-;/m0./s1
Formula Weight 248.1137
Percent Purity >95%
Chemical Name or Material (2S,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Pilocarpic Acid Sodium Salt)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.