missing translation for 'onlineSavingsMsg'
Learn More

Pilocarpine Hydrochloride, TRC

Product Code. 30411364
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411364 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411364 Supplier Toronto Research Chemicals Supplier No. TRCP4415005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pilocarpine Hydrochloride
CAS 54-71-7
Molecular Formula C11 H16 N2 O2 . Cl H
Quantity 5 g
Synonym Pilocarpine Hydrochloride, (3S,4R)-3-Ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-2(3H)-furanone monohydrochloride, (3S-cis)-3-Ethyldihydro-4-[(1-methyl-1H-imidazol-5-yl)methyl]-2(3H)-furanone monohydrochloride, Pilocarpine monohydrochloride, (+)-Pilocarpine hydrochloride, Adsorbocarpine, Almocarpine, Isopto Carpine, NSC 5746, Pilocar SMP, Pilocarpal, Pilocarpine chloride, Pilocarpine hydrochloride, Pilocarpine muriate, Pilocarpinium chloride, Pilocel, Pilovisc, Salagen, Vistacarpin N
InChI Formula InChI=1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10-;/m0./s1
SMILES Cl.CC[C@H]1[C@@H](Cc2cncn2C)COC1=O
IUPAC Name (3S,4R)-3-ethyl-4-[(3-methylimidazol-4-yl)methyl]oxolan-2-one;hydrochloride
Molecular Weight (g/mol) 244.72
Formula Weight 244.0979
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.