missing translation for 'onlineSavingsMsg'
Learn More

Pimobendan, MedChemExpress

Product Code. 16460818
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460818 5 mg 5mg
16438418 10 mg 10mg
16404307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16460818 Supplier Medchem Express Supplier No. HYB0204
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pimobendan (UD-CG115) is a selective inhibitor of PDE3 with IC50 of 0.32 μM.

Pimobendan (UD-CG115) is a selective inhibitor of PDE3 with IC50 of 0.32 μM.

Specifications

Chemical Name or Material Pimobendan
CAS 74150-27-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H18N4O2
Quantity 5 mg
Synonym UD-CG115
Solubility Information DMSO : 50 mg/mL (149.53 mM; Need ultrasonic)
SMILES O=C1CC(C)C(C2=CC=C3N=C(C4=CC=C(OC)C=C4)NC3=C2)=NN1
Molecular Weight (g/mol) 334.37
Formula Weight 334.37
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.