missing translation for 'onlineSavingsMsg'
Learn More

Pinacolborane, 97%

Product Code. 11318506
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318506 5 g 5g
11328506 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318506 Supplier Thermo Scientific Alfa Aesar Supplier No. L17558.06
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 25015-63-8 | C6H13BO2 | 127.98 g/mol

Pinacolborane is used as a precursor of boronic esters by hydroboration or coupling reactions. It acts as a monofunctional hydroborating agent. It is used in the synthesis of unsymmetrical biaryls through aromatic C-H borylation-cross-coupling sequence. It is involved in the preparation of 4,4,5,5-tetramethyl-2-thiophen-2-yl-[1,3,2]dioxaborolane by reacting with 2-iodo-thiophene. Further, it is used to prepare vinylboronates by palladium-catalyzed coupling with alkenyl triflates and iodides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pinacolborane is used as a precursor of boronic esters by hydroboration or coupling reactions. It acts as a monofunctional hydroborating agent. It is used in the synthesis of unsymmetrical biaryls through aromatic C-H borylation-cross-coupling sequence. It is involved in the preparation of 4,4,5,5-tetramethyl-2-thiophen-2-yl-[1,3,2]dioxaborolane by reacting with 2-iodo-thiophene. Further, it is used to prepare vinylboronates by palladium-catalyzed coupling with alkenyl triflates and iodides.

Solubility
Miscible with ether, dichloromethane, tetrahydrofuran and organic solvents.

Notes
Air sensitive and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids, water, bases and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25015-63-8
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.98
MDL Number MFCD00674030
InChI Key UCFSYHMCKWNKAH-UHFFFAOYSA-N
Synonym pinacolborane, 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, pinb-te, pubchem12688, acmc-209ggf, pinacolborane 5g, ksc489k6t, ucfsyhmckwnkah-uhfffaoysa-n
PubChem CID 6364989
IUPAC Name 4,4,5,5-tetramethyl-1,3,2$l^{2}-dioxaborolane
SMILES CC1(C)OBOC1(C)C

Specifications

Density 0.882
Boiling Point 42°C to 43°C (50 mmHg)
Flash Point 5°C (41°F)
Refractive Index 1.397
Quantity 5 g
UN Number UN3399
Beilstein 7802871
Sensitivity Air sensitive
Solubility Information Miscible with ether,dichloromethane,tetrahydrofuran and organic solvents.
Formula Weight 127.98
Percent Purity 97%
Chemical Name or Material Pinacolborane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.