missing translation for 'onlineSavingsMsg'
Learn More

Pinacolborane, TRC

Product Code. 30412308
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412308 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412308 Supplier Toronto Research Chemicals Supplier No. TRCP45750025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25015-63-8
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.98
Synonym 4,4,5,5-Tetramethyl-1,3,2-dioxaborolane, Pinacolatoborane
IUPAC Name 4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OBOC1(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H13BO2/c1-5(2)6(3,4)9-7-8-5/h7H,1-4H3
Formula Weight 128.1009
Chemical Name or Material Pinacolborane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.