missing translation for 'onlineSavingsMsg'
Learn More

Pinacolone, 97%, Thermo Scientific Chemicals

Product Code. 11484023
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484023 500 g 500g
11474023 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11484023 Supplier Thermo Scientific Alfa Aesar Supplier No. A10289.36
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.

Solubility
Miscible with water, chloroform, ethanol, ether, acetone and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-97-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008846
InChI Key PJGSXYOJTGTZAV-UHFFFAOYSA-N
Synonym pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone
PubChem CID 6416
IUPAC Name 3,3-dimethylbutan-2-one
SMILES CC(=O)C(C)(C)C

Specifications

Melting Point -53°C
Density 0.803
Boiling Point 105°C to 106°C
Flash Point 5°C (41°F)
Assay Percent Range 97%
Linear Formula (CH3)3CC(O)CH3
Refractive Index 1.397
Quantity 500 g
UN Number UN1224
Merck Index 14,7440
Solubility Information Miscible with water,chloroform,ethanol,ether,acetone and carbon tetrachloride.
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material Pinacolone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.