missing translation for 'onlineSavingsMsg'
Learn More

alpha-Pinene, 97%, stabilized with alpha-Tocopherol

Product Code. 10397530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
Product Code. Quantity Packaging unitSize
10397530 250 g Glass bottle 250g
10701441 5 g Glass bottle 5g
10584634 1 kg Glass bottle 1kg
3 options
This item is not returnable. View return policy

Product Code. 10397530

Brand: Thermo Scientific Acros 164362500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-56-8 | C10H16 | 136.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-56-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001339
InChI Key GRWFGVWFFZKLTI-UHFFFAOYSA-N
Synonym alpha-pinene, 2-pinene, .alpha.-pinene, acintene a, pinene isomer, 2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, pinene, alpha, pin-2 3-ene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl, +-a-pinene
PubChem CID 6654
ChEBI CHEBI:36740
IUPAC Name 4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES CC1=CCC2CC1C2(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -55.0°C
Color Colorless
Density 0.8500g/mL
Boiling Point 154.0°C to 157.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 250 g
Merck Index 15, 7556
Specific Gravity 0.85
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,chloroform and ether
Stabilizer 0.8% alpha-tocopherol
Viscosity <7 mm2/s (40°C)
Formula Weight 136.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material α-Pinene, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.