missing translation for 'onlineSavingsMsg'
Learn More

(+)-Pinoresinol (>80% ee), TRC

Product Code. 17908394
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17908394 5 mg 5mg
17927324 50 mg 50mg
17908404 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17908394 Supplier Toronto Research Chemicals Supplier No. TRCP4688905MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-36-5
Molecular Formula C20 H22 O6
Molecular Weight (g/mol) 358.38
Synonym 4,4'-((1S,3aR,4S,6aR)-Hexahydrofuro[3,4-c]furan-1,4-diyl)bis(2-methoxyphenol), Pinoresinol, (+)-Pinoresinol, Phenol, 4,4'-[(1S,3aR,4S,6aR)-tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis[2-methoxy-, Phenol, 4,4'-(tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl)bis[2-methoxy-, [1S-(1α,3aα,4α,6aα)]-, Phenol, 4,4'-(tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl)bis[2-methoxy-, stereoisomer (8CI), Pinoresinol (6CI,7CI), 1H,3H-Furo[3,4-c]furan, phenol deriv., (+)-Pinoresinol, 4,4'-[(1S,3aR,4S,6aR)-Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis[2-methoxyphenol], NSC 35444, d-Pinoresinol, 1H,3H-Furo[3,4-c]furan Phenol Derivative, Pinoresinol
IUPAC Name 4-[(3S,3aR,6S,6aR)-6-(4-hydroxy-3-methoxyphenyl)-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-3-yl]-2-methoxyphenol
SMILES COc1cc(ccc1O)[C@H]2OC[C@H]3[C@@H]2CO[C@@H]3c4ccc(O)c(OC)c4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H22O6/c1-23-17-7-11(3-5-15(17)21)19-13-9-26-20(14(13)10-25-19)12-4-6-16(22)18(8-12)24-2/h3-8,13-14,19-22H,9-10H2,1-2H3/t13-,14-,19+,20+/m0/s1
Formula Weight 358.1416
Percent Purity >95%
Chemical Name or Material (+)-Pinoresinol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.