missing translation for 'onlineSavingsMsg'
Learn More

Pinoresinol Diglucoside, TRC

Product Code. 30409981
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409981 10 mg 10mg
30417214 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30409981 Supplier Toronto Research Chemicals Supplier No. TRCP46890010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pinoresinol diglucoside
CAS 63902-38-5
Molecular Formula C32 H42 O16
Quantity 10 mg
Synonym β-D-Glucopyranoside','[(1S,3aR,4S,6aR)-tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2-methoxy-4,1-phenylene) bis-, β-D-Glucopyranoside','(tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl)bis(2-methoxy-4,1-phenylene) bis-','[1S-(1α,3aα,4α,6aα)]-, 1H,3H-Furo[3,4-c]furan','β-D-glucopyranoside deriv., (+)-Pinoresinol di-O-β-D-glucopyranoside, (+)-Pinoresinol di-β-D-glucopyranoside, Pinoresinol di-β-D-glucoside, Pinoresinol diglucoside, [(1S,3aR,4S,6aR)-Tetrahydro-1H,3H-furo[3,4-c]furan-1,4-diyl]bis(2-methoxy-4,1-phenylene) bis-β-D-glucopyranoside
InChI Formula InChI=1S/C32H42O16/c1-41-19-7-13(3-5-17(19)45-31-27(39)25(37)23(35)21(9-33)47-31)29-15-11-44-30(16(15)12-43-29)14-4-6-18(20(8-14)42-2)46-32-28(40)26(38)24(36)22(10-34)48-32/h3-8,15-16,21-40H,9-12H2,1-2H3/t15-,16-,21+,22+,23+,24+,25-,26-,27+,28+,29+,30+,31+,32+/m0/s1
SMILES COc1cc(ccc1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]3OC[C@H]4[C@@H]3CO[C@@H]4c5ccc(O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c(OC)c5
IUPAC Name (2S,3R,4S,5S,6R)-2-[4-[(3S,3aR,6S,6aR)-6-[3-methoxy-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan-3-yl]-2-methoxyphenoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 682.67
Formula Weight 682.2473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.