missing translation for 'onlineSavingsMsg'
Learn More

Pioglitazone hydrochloride, 98%

Product Code. 15486978
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486978 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15486978 Supplier Thermo Scientific Alfa Aesar Supplier No. H60507.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112529-15-4 | C19H21ClN2O3S | 392.898 g/mol

Pioglitazone hydrochloride exhibits antiinflammatory and antiarteriosclerotic effects. It is used to prevent L-NAME-induced coronary inflammation and arteriosclerosis and to suppress increased TNF-α mRNA produced by aspirin-induced gastric mucosal injury. Pioglitazone Hydrochloride is an activator of PPAR γ. It is a PPARγ agonist and thiazolidinedione (TZD) anti-diabetic. Pioglitazone is a selective agonist of the nuclear receptor peroxisome proliferator-activated receptor γ (PPAR-γ) and to a lesser extent PPAR-α.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pioglitazone hydrochloride exhibits antiinflammatory and antiarteriosclerotic effects. It is used to prevent L-NAME-induced coronary inflammation and arteriosclerosis and to suppress increased TNF-α mRNA produced by aspirin-ind
TRUSTED_SUSTAINABILITY

Specifications

CAS 112529-15-4
Molecular Formula C19H21ClN2O3S
MDL Number MFCD04975446
Quantity 250 mg
Synonym pioglitazone hydrochloride, pioglitazone hcl, 5-4-2-5-ethylpyridin-2-yl ethoxy benzyl thiazolidine-2,4-dione hydrochloride, pioditazone hydrochloride, 5-4-2-5-ethyl-2-pyridinyl ethoxy phenyl methyl-2,4-thiazolidinedione hydrochloride, +--5-p-2-5-ethyl-2-pyridyl ethoxy benzyl-2,4-thiazolidinedione monohydrochloride, dsstox_cid_24203, dsstox_rid_80116, dsstox_gsid_44203
Solubility Information Soluble in water (<1mg/ml at 25°C),DMF,methanol,ethanol (4mg/ml at 25°C),and DMSO (79mg/ml at 25°C).
InChI Key GHUUBYQTCDQWRA-UHFFFAOYSA-N
SMILES CCC1=CN=C(C=C1)CCOC2=CC=C(C=C2)CC3C(=O)NC(=O)S3.Cl
IUPAC Name 5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride
Molecular Weight (g/mol) 392.898
PubChem CID 60560
Formula Weight 392.9
Percent Purity 98%
Chemical Name or Material Pioglitazone hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.