Learn More
Invitrogen™ Human PIP4K2A, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIP4K2A recombinant human protein (full length) was expressed in insect cells. PIP4K2A recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for PIP4K2A is NP_005019.2.
- Validated for use with the Adapta™ assay
- Available in multiple pack sizes for convenient use
Specifications
Specifications
| Accession Number | NP_005019.2 |
| For Use With (Application) | Kinase Assay |
| Molecular Weight (g/mol) | 72.5 kDa |
| Quantity | 1 mg |
| KinaseFamily | PI3 Kinase |
| Protein Length | Full Length |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | GST Tag |
| Content And Storage | Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.