missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pipe Markers (On Liner) - Standard Legends

Product Code. 15319015 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
355 mm
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
15319015 355 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15319015 Supplier Brady™ Supplier No. N006986
  • ONLINE EXCLUSIVE PRICE
    17.05 €
    16.31 € / Pack of 3
    Save 0.74 €
    4% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required.

Specifications

Description Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Acids and Alkalis - Acid
Adhesive Type Permanent Acrylic
Color White on Purple
Finish Gloss
Format Card
Material Polyester
Product Type Pipe Marker
Display Text Acid
For Use With (Application) Pipe and valve marking
Labels per Sheet 3
Languages English
No. per Pack 3 Markers/Card
Quantity 3 Pk.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness (English) 0.0039 in.
Thickness (Metric) 0.099 mm
Width (English) 13.98 in.
Width (Metric) 355 mm
Height (English) 1.46 in.
Height (Metric) 37 mm
Alternate Part Number ACID 355X37CARD-T1-P21
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.