missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pipemarkers on a Card

Product Code. 15399115 Shop All Brady Products
Change view
Click to view available options
Display Text:
Argon
Bio gas
CO2
Carbon dioxide
Condensate
Cooling Gas
Freon
Freon 11
Gas
Helium
Inert Gas
LPG
Mixed Gas
Natural gas
Nitrogen
Recycled Gas
Return
Color:
Black on Ochre
Unit Size:
Pack of 5
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Color Unit Size
15399115 Recycled Gas Black on Ochre -
15359025 Argon Black on Ochre Pack of 5
15319095 Carbon dioxide Black on Ochre -
15339045 Bio gas Black on Ochre -
15379055 Condensate Black on Ochre -
15309055 CO2 Black on Ochre Pack of 5
15329105 Cooling Gas Black on Ochre -
15329075 Freon Black on Ochre -
15389085 Gas Black on Ochre Pack of 5
15389075 Freon 11 Black on Ochre -
15379125 Helium Black on Ochre -
15369105 Inert Gas Black on Ochre -
15329125 LPG Black on Ochre Pack of 5
15399105 Mixed Gas Black on Ochre Pack of 5
15329115 Natural gas Black on Ochre -
15359035 Nitrogen Black on Ochre Pack of 5
15309135 Return Black on Ochre Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 15399115 Supplier Brady™ Supplier No. N007616
  • ONLINE EXCLUSIVE PRICE
    13.70 €
    13.05 € / Pack of 5
    Save 0.65 €
    4% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Regulations 1271/2008, 790/2009 and 286/2001
Description Individual Pipe Markers on a Card with die-cut arrowheads, with pictograms - Gas - Recycled gas
Adhesive Type Permanent Rubber, Permanent, Pressure-sensitive
Color Black on Ochre
Display Warning
Format Card
Material Polyester
Print Technology Thermal Transfer
Product Type Pipe Marker
Display Text Recycled Gas
For Use With (Application) Pipe and valve marking
Labels per Sheet 5
Languages English
No. per Pack 5 Markers/Card
Quantity 5 Pk.
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Thickness (English) 0.0049 in.
Thickness (Metric) 0.124 mm
Dimensions (L x W) 12 x 150 mm
Width (English) 5.91 in.
Width (Metric) 150 mm
Height (English) 0.47 in.
Height (Metric) 12 mm
Height (Metric) Character 8 mm
Alternate Part Number RECYCLED GAS 150X12CARD-T1-P15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.