missing translation for 'onlineSavingsMsg'
Learn More

Piperazine hexahydrate, 98%

Product Code. 10317020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10317020 100 g Plastic bottle 100g
10645762 500 g Plastic bottle 500g
10795611 1 kg Plastic bottle 1kg
13418358 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10317020 Supplier Thermo Scientific Acros Supplier No. 131301000
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-63-2 | C4H10N2·6H2O | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-63-2
Molecular Formula C4H10N2·6H2O
Molecular Weight (g/mol) 194.23
MDL Number MFCD00149389
InChI Key AVRVZRUEXIEGMP-UHFFFAOYSA-N
Synonym piperazine hexahydrate, arthriticine, piperazine, hexahydrate, vermisol, parid, unii-p3m07b8u64, usaf a-3803, diethylenediamine hexahydrate, vermyl tn
PubChem CID 120181
IUPAC Name piperazine;hexahydrate
SMILES C1CNCCN1.O.O.O.O.O.O

Specifications

Melting Point 41.0°C to 45.0°C
Color White
Boiling Point 145.0°C to 156.0°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 23,4
Merck Index 14,7464
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol,practically insoluble in ether
Formula Weight 194.23
Percent Purity 98%
Physical Form Adhering Crystals
Chemical Name or Material Piperazine hexahydrate, 98%
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • Piperazine hexahydrate
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Faroangivelser
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H314-Causes severe skin burns and eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.