missing translation for 'onlineSavingsMsg'
Learn More

Piperazine, TRC

Product Code. 30132568
Change view
Click to view available options
Quantity:
2.5 g
25 g
500 g
Unit Size:
2.50g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132568 500 g 500g
30004345 25 g 25g
30003678 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132568 Supplier Toronto Research Chemicals Supplier No. TRCP479900500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-85-0
Molecular Formula C4 H10 N2
Molecular Weight (g/mol) 86.14
Synonym Piperazine, 1,4-Diazacyclohexane, 1,4-Piperazine, Antiren, Diethylenediamine, Dispermine, Eraverm, Hexahydropyrazine, Lumbrical, NSC 474, Piperazidine, Pipersol, Pyrazine hexahydride, Pyrazine, hexahydro-, Tetrahydro-1,4-diazine, Uvilon, Vermex, Worm-A-Ton, Wurmirazin
IUPAC Name piperazine
SMILES C1CNCCN1

Specifications

Quantity 500 g
InChI Formula InChI=1S/C4H10N2/c1-2-6-4-3-5-1/h5-6H,1-4H2
Formula Weight 86.0844
Percent Purity >95%
Chemical Name or Material Piperazine
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.