missing translation for 'onlineSavingsMsg'
Learn More

piperidine-4-carboxylic acid, Maybridge™

Product Code. 10225284
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10225284

Brand: Thermo Scientific Maybridge SB00736EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 498-94-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006004
InChI Key SRJOCJYGOFTFLH-UHFFFAOYSA-N
Synonym isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
PubChem CID 3773
IUPAC Name piperidine-4-carboxylic acid
SMILES OC(=O)C1CCNCC1

Specifications

Assay Percent Range 97+%
Quantity 25 g
Formula Weight 129.16
Percent Purity 97%
Chemical Name or Material Isonipecotic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.