missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Piperidine, 99%

Product Code. 11449663
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449663 500 mL 500mL
11404030 100 mL 100mL
11459663 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 11449663 Supplier Thermo Scientific Chemicals Supplier No. A12442.AP
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

CAS: 110-89-4 | C5H11N | 85.15 g/mol

Extensively finds applications as a base, a nucleophile and a solvent. Used for the production of dipiperidinyl dithiuram tetrasulfide, which is used as a rubber vulcanization accelerator. It is commonly used in solid phase peptide synthesis (F-MOC Chemistry) and in chemical degradation reactions (sequencing of DNA). Used in making rubber, curing epoxy resins and in fine chemicals, and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Piperidine, 99% 500mL Plastic bottle CAS No. 110-89-4
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-89-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
InChI Key NQRYJNQNLNOLGT-UHFFFAOYSA-N
IUPAC Name piperidine
SMILES C1CCNCC1

Specifications

Melting Point -11°C
Boiling Point 106°C (literature)
Packaging Plastic
Quantity 500 mL
Percent Purity 99%
Chemical Name or Material Piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.