missing translation for 'onlineSavingsMsg'
Learn More

Piperidino(4-piperidinyl)methanone, ≥97%, Thermo Scientific™

Product Code. 10535733
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10535733 250 mg Amber Glass Bottle 250mg
10041692 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10535733 Supplier Thermo Scientific Maybridge Supplier No. MO01170CB
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 63214-58-4
Molecular Formula C11H21N2O
Molecular Weight (g/mol) 197.30
MDL Number MFCD03372500
InChI Key JFHHGGAQABVJIF-UHFFFAOYSA-O
Synonym 1-piperidin-4-ylcarbonyl piperidine, piperidin-1-yl piperidin-4-yl methanone, piperidino 4-piperidinyl methanone, 1-piperidine-4-carbonyl piperidine, 4-piperidine-1-carbonyl piperidine, piperidin-4-yl-piperidin-1-yl-methanone, methanone,1-piperidinyl-4-piperidinyl, 1-piperidin-4-yl carbonyl piperidine, piperidine, 1-4-piperidinylcarbonyl
PubChem CID 2794688
SMILES O=C(C1CC[NH2+]CC1)N1CCCCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name piperidin-1-yl(piperidin-4-yl)methanone
Formula Weight 196.29
Percent Purity 97%
Chemical Name or Material Piperidino(4-piperidinyl)methanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.