missing translation for 'onlineSavingsMsg'
Learn More

Piperine, 98%

Product Code. 11425613
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425613 100 g 100g
11405613 5 g 5g
11415613 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425613 Supplier Thermo Scientific Alfa Aesar Supplier No. A13510.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-62-2 | C17H19NO3 | 285.34 g/mol

Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.

Solubility
Soluble in water(0.04g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-62-2
Molecular Formula C17H19NO3
Molecular Weight (g/mol) 285.34
MDL Number MFCD00005839
InChI Key MXXWOMGUGJBKIW-YPCIICBESA-N
Synonym piperine, 1-piperoylpiperidine, piperin, bioperine, piperoylpiperidine, e,e-1-piperoylpiperidine, n-e,e-piperoyl piperidine, piperidine, 1-piperoyl-, e,e, unii-u71xl721qk, fema no. 2909
PubChem CID 638024
ChEBI CHEBI:28821
IUPAC Name (2E,4E)-5-(1,3-benzodioxol-5-yl)-1-piperidin-1-ylpenta-2,4-dien-1-one
SMILES O=C(\C=C\C=C\C1=CC=C2OCOC2=C1)N1CCCCC1

Specifications

Melting Point 128°C to 133°C
Quantity 100 g
Beilstein 90741
Merck Index 14,7472
Solubility Information Soluble in water(0.04g/L).
Formula Weight 285.34
Percent Purity 98%
Chemical Name or Material Piperine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.